1-Carbamoylmethylpiperidine-4-carboxylic acid ethyl ester ≥95%
Supplier: Matrix Scientific
058833-1G
058833-5G
101915-972EA
380
USD
101915-972
101915-974
1-Carbamoylmethylpiperidine-4-carboxylic acid ethyl ester ≥95%
1-Carbamoylmethylpiperidine-4-carboxylic acid ethyl ester
Hazard: Irritant
Formula:
C₁₀H₁₈N₂O₃ |
MDL Number:
MFCD09837321 CAS Number: 40479-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...