1-tert-Butyloxycarbonylpiperazine-4-acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(cyanomethyl)piperazine-1-carboxylate

058829-1G 058829-5G 058829-10G
101915-960EA 378.46 USD
101915-960 101915-962 101915-964
1-tert-Butyloxycarbonylpiperazine-4-acetonitrile ≥95%
1-tert-Butyloxycarbonylpiperazine-4-acetonitrile

Hazard: Irritant

Formula: C₁₁H₁₉N₃O₂
MW: 225.29 g/mol
MDL Number: MFCD06795916
CAS Number: 77290-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR