tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate ≥94%
Supplier: Matrix Scientific
Synonyms:
1-tert-Butyloxycarbonyl-3-bromo-4-oxopiperidine, tert-Butyl 3-bromo-4-oxopiperidine-1-carboxylate
058827-1G
058827-5G
101915-954EA
18.46
USD
101915-954
101915-956
tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate ≥94%
tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₆BrNO₃ MW: 278.1429 g/mol |
MDL Number:
MFCD06738530 CAS Number: 188869-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...