tert-Butyl-6-fluoropyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-fluoropicolinate

058819-1G 058819-5G
101915-928EA 407.69 USD
101915-928 101915-930
tert-Butyl-6-fluoropyridine-2-carboxylate ≥95%
tert-Butyl-6-fluoropyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₂FNO₂
MW: 197.21 g/mol
MDL Number: MFCD10568316
CAS Number: 1053656-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR