3-(tert-Butoxycarbonylethoxycarbonylmethylamino)propionic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-[(N-tert-butoxycarbonylethoxycarbonyl)methylamino]propanoate

058818-1G 058818-5G
101915-924EA 523.08 USD
101915-924 101915-926
3-(tert-Butoxycarbonylethoxycarbonylmethylamino)propionic acid ethyl ester ≥95%
3-(tert-Butoxycarbonylethoxycarbonylmethylamino)propionic acid ethyl ester

Hazard: Irritant

Formula: C₁₄H₂₅NO₆
MW: 303.36 g/mol
MDL Number: MFCD15142792
CAS Number: 146256-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR