5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine

058816-1G 058816-5G 058816-250MG
101915-918EA 30.77 USD
101915-918 101915-920 102551-546
5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine ≥95%
5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine

Hazard: Irritant

Formula: C₅H₃BrF₃N₃
MDL Number: MFCD09261254
CAS Number: 935534-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR