5-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-4-(trifluoromethyl)pyridin-2-ol

058813-1G 058813-5G
101915-908EA 181.54 USD
101915-908 101915-910
5-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine ≥98%
5-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃BrF₃NO
MW: 241.9933 g/mol
MDL Number: MFCD11227189
CAS Number: 109919-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR