5-Bromo-1,3,4-thiadiazol-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-1,3,4-thiadiazole, 1,3,4-Thiadiazol-2-amine, 5-bromo-, 5-bromo-1,3,4-thiadiazol-2-amine

058811-5G 058811-25G
101915-902EA 46.15 USD
101915-902 101915-904
5-Bromo-1,3,4-thiadiazol-2-amine ≥95%
5-Bromo-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₂H₂BrN₃S
MW: 180.02 g/mol
MDL Number: MFCD00464325
CAS Number: 37566-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR