2-Bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone ≥95%

Supplier: Matrix Scientific

058809-1G 058809-5G 058809-10G
101915-892EA 152.31 USD
101915-892 101915-894 101915-896
2-Bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone ≥95%
2-Bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone

Hazard: Irritant

Formula: C₁₁H₈BrNOS
MW: 282.17 g/mol
MDL Number: MFCD02677697
CAS Number: 306935-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR