5-Bromopyridine-2-carbonyl chloride ≥95%

Supplier: Matrix Scientific

058807-1G 058807-5G
101915-886EA 238.46 USD
101915-886 101915-888
5-Bromopyridine-2-carbonyl chloride ≥95%
5-Bromopyridine-2-carbonyl chloride

Hazard: Irritant

Formula: C₆H₃BrClNO
MDL Number: MFCD07368290
CAS Number: 137178-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR