2-(4-Bromophenyl)thioacetamide ≥95%

Supplier: Matrix Scientific

058806-1G 058806-5G
101915-882EA 580 USD
101915-882 101915-884
2-(4-Bromophenyl)thioacetamide ≥95%
2-(4-Bromophenyl)thioacetamide

Hazard: Irritant

Formula: C₈H₈BrNS
MDL Number: MFCD09809469
CAS Number: 147111-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR