2-(4-Bromophenylsulfanyl)-3-chloro-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

058804-1G 058804-5G
101915-876EA 524.62 USD
101915-876 101915-878
2-(4-Bromophenylsulfanyl)-3-chloro-5-(trifluoromethyl)pyridine ≥95%
2-(4-Bromophenylsulfanyl)-3-chloro-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₆BrClF₃NS
MW: 368.61 g/mol
MDL Number: MFCD09864945
CAS Number: 1053658-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR