2-Bromo-4-phenylnicotinic acid ≥95%

Supplier: Matrix Scientific

058802-1G 058802-5G
101915-870EA 669.23 USD
101915-870 101915-872
2-Bromo-4-phenylnicotinic acid ≥95%
2-Bromo-4-phenylnicotinic acid

Hazard: Irritant

Formula: C₁₂H₈BrNO₂
MDL Number: MFCD18384842
CAS Number: 147078-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR