N'-[1-(4-Bromophenyl)meth-(E)-ylidene]-N,N-dimethylethane-1,2-diamine ≥95%

Supplier: Matrix Scientific

058799-500MG 058799-1G
101915-862EA 553.85 USD
101915-862 101915-864
N'-[1-(4-Bromophenyl)meth-(E)-ylidene]-N,N-dimethylethane-1,2-diamine ≥95%
N'-[1-(4-Bromophenyl)meth-(E)-ylidene]-N,N-dimethylethane-1,2-diamine

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂
MW: 255.16 g/mol
MDL Number: MFCD15142809
CAS Number: 91345-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR