1-(4-Bromo-phenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Bromophenyl)-1H-pyrazole

058798-1G 058798-5G
101915-856EA 61.54 USD
101915-856 101915-858
1-(4-Bromo-phenyl)-1H-pyrazole ≥95%
1-(4-Bromo-phenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₇BrN₂
MW: 223.08 g/mol
MDL Number: MFCD08059318
CAS Number: 13788-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR