1-(3-Bromophenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

058797-1G 058797-5G 058797-10G
101915-850EA 263.08 USD
101915-850 101915-852 101915-854
1-(3-Bromophenyl)-1H-pyrazole ≥95%
1-(3-Bromophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD06797472
CAS Number: 294877-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR