Ethyl-6-(bromomethyl)nicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-(bromomethyl)nicotinate

058785-500MG 058785-1G
101915-810EA 526.15 USD
101915-810 101915-812
Ethyl-6-(bromomethyl)nicotinate ≥95%
Ethyl-6-(bromomethyl)nicotinate

Hazard: Irritant

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
MDL Number: MFCD10566281
CAS Number: 178264-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR