6-Bromomethyl-nicotinic acid methyl ester ≥90%
Supplier: Matrix Scientific
058783-1G
058783-5G
101915-802EA
495.38
USD
101915-802
101915-804
6-Bromomethyl-nicotinic acid methyl ester ≥90%
6-Bromomethyl-nicotinic acid methyl ester
Hazard: Irritant
Formula:
C₈H₈BrNO₂ |
MDL Number:
MFCD10566280 CAS Number: 131803-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...