5-Bromo-3-methylisothiazole ≥95%

Supplier: Matrix Scientific

058780-1G 058780-5G
101915-790EA 584.62 USD
101915-790 101915-792
5-Bromo-3-methylisothiazole ≥95%
5-Bromo-3-methylisothiazole

Hazard: Irritant

Formula: C₄H₄BrNS
MW: 178.06 g/mol
MDL Number: MFCD01318949
CAS Number: 20493-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR