5-Bromo-6-hydroxynicotinic acid ≥95%

Supplier: Matrix Scientific

058779-1G 058779-5G
101915-786EA 23.08 USD
101915-786 101915-788
5-Bromo-6-hydroxynicotinic acid ≥95%
5-Bromo-6-hydroxynicotinic acid

Hazard: Irritant

Formula: C₆H₄BrNO₃
MDL Number: MFCD08235173
CAS Number: 41668-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR