1'-(5-Bromo-6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine hydrobromide ≥95%

Supplier: Matrix Scientific

058775-1G 058775-5G
101915-772EA 523.08 USD
101915-772 101915-774
1'-(5-Bromo-6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine hydrobromide ≥95%
1'-(5-Bromo-6-chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine hydrobromide

Hazard: Irritant

Formula: C₁₀H₁₁Br₂ClF₃N₃
MDL Number: MFCD11052384
CAS Number: 1208081-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR