5-Bromo-2-chloromethyl-1H-benzoimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-(chloromethyl)-1H-benzo[d]imidazole

058774-1G 058774-5G 058774-10G
101915-766EA 220 USD
101915-766 101915-768 101915-770
5-Bromo-2-chloromethyl-1H-benzoimidazole ≥95%
5-Bromo-2-chloromethyl-1H-benzoimidazole

Hazard: Irritant

Formula: C₈H₆BrClN₂
MW: 245.51 g/mol
MDL Number: MFCD00457491
CAS Number: 1740-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR