(5-Bromo-benzo[b]thiophen-2-yl)methanol ≥95%

Supplier: Matrix Scientific

058769-500MG 058769-1G 058769-5G
101915-746EA 453.85 USD
101915-746 101915-748 101915-750
(5-Bromo-benzo[b]thiophen-2-yl)methanol ≥95%
(5-Bromo-benzo[b]thiophen-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇BrOS
MDL Number: MFCD12032279
CAS Number: 13771-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR