6-Bromobenzo[b]thiophene-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

058768-500MG 058768-1G 058768-5G
101915-740EA 293.85 USD
101915-740 101915-742 101915-744
6-Bromobenzo[b]thiophene-2-carbaldehyde ≥95%
6-Bromobenzo[b]thiophene-2-carbaldehyde

Hazard: Irritant

Formula: C₉H₅BrOS
MW: 241.11 g/mol
MDL Number: MFCD09834665
CAS Number: 19075-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR