(2,4-Bis(trifluoromethylphenyl))oxoacetic acid ethyl ester, Technical Grade

Supplier: Matrix Scientific

058765-500MG 058765-1G 058765-5G
101915-724EA 466.15 USD
101915-724 101915-726 101915-728
(2,4-Bis(trifluoromethylphenyl))oxoacetic acid ethyl ester, Technical Grade
(2,4-Bis(trifluoromethylphenyl))oxoacetic acid ethyl ester

Hazard: Irritant

Formula: C₁₂H₈F₆O₃
MW: 314.19 g/mol
MDL Number: MFCD01934869
CAS Number: 306936-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR