3-Benzyloxy-4-methylphenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Benzyloxy)-4-methylaniline

058764-1G 058764-5G 058764-10G
101915-718EA 393.85 USD
101915-718 101915-720 101915-722
3-Benzyloxy-4-methylphenylamine ≥95%
3-Benzyloxy-4-methylphenylamine

Hazard: Irritant

Formula: C₁₄H₁₅NO
MW: 213.28 g/mol
MDL Number: MFCD04973042
CAS Number: 219492-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR