1-Benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

058760-10MG 058760-50MG
101915-706EA 233.85 USD
101915-706 101915-708
1-Benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylic acid ethyl ester ≥95%
1-Benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₄H₁₅NO₄
MDL Number: MFCD00179167
CAS Number: 4450-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR