N'-[1-Benzylamino-1-(2,2,2-trifluoroethylamino)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058756-500MG 058756-1G
101915-690EA 284.62 USD
101915-690 101915-692
N'-[1-Benzylamino-1-(2,2,2-trifluoroethylamino)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Benzylamino-1-(2,2,2-trifluoroethylamino)methylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₅H₂₁F₃N₄O₂
MW: 346.36 g/mol
MDL Number: MFCD10568243
CAS Number: 1053657-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR