N'-[1-Amino-1-(3-(trifluoromethyl)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058749-500MG
058749-1G
101915-666EA
284.62
USD
101915-666
101915-668
N'-[1-Amino-1-(3-(trifluoromethyl)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(3-(trifluoromethyl)phenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₆F₃N₃O₂ MW: 303.29 g/mol |
MDL Number:
MFCD10568174 CAS Number: 1053655-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...