2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-amino-1,3-thiazole-4-carboxylate hydrobromide, Ethyl 2-aminothiazole-4-carboxylate hydrobromide

058746-500MG 058746-1G 058746-5G
101915-652EA 378.46 USD
101915-652 101915-654 101915-656
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide ≥95%
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide

Hazard: Irritant

Formula: C₆H₉BrN₂O₂S
MW: 253.12 g/mol
MDL Number: MFCD06653384
CAS Number: 127942-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR