2-Aminothiazole-5-carboxylic acid ethyl ester hydrochloride ≥95%

Supplier: Matrix Scientific

058745-1G 058745-5G
101915-648EA 495.38 USD
101915-648 101915-650
2-Aminothiazole-5-carboxylic acid ethyl ester hydrochloride ≥95%
2-Aminothiazole-5-carboxylic acid ethyl ester hydrochloride

Hazard: Irritant

Formula: C₆H₉ClN₂O₂S
MDL Number: MFCD11035752
CAS Number: 162849-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR