N'-[1-Amino-1-p-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058744-500MG
058744-1G
101915-644EA
284.62
USD
101915-644
101915-646
N'-[1-Amino-1-p-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-p-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₉N₃O₂ MW: 249.32 g/mol |
MDL Number:
MFCD10568182 CAS Number: 1053655-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...