N'-[1-Amino-2-p-tolylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058743-500MG
058743-1G
101915-640EA
284.62
USD
101915-640
101915-642
N'-[1-Amino-2-p-tolylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-p-tolylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₄H₂₁N₃O₂ MW: 263.34 g/mol |
MDL Number:
MFCD10568177 CAS Number: 159016-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...