N'-[1-Aminopropylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058742-500MG
058742-1G
101915-636EA
284.62
USD
101915-636
101915-638
N'-[1-Aminopropylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Aminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₈H₁₇N₃O₂ MW: 187.24 g/mol |
MDL Number:
MFCD10568290 CAS Number: 159016-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...