N'-[1-Amino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058738-500MG
058738-1G
101915-624EA
284.62
USD
101915-624
101915-626
N'-[1-Amino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₉N₃O₂ MW: 249.32 g/mol |
MDL Number:
MFCD10568176 CAS Number: 159016-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...