N'-[1-Amino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058737-500MG 058737-1G
101915-620EA 284.62 USD
101915-620 101915-622
N'-[1-Amino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₃
MW: 265.31 g/mol
MDL Number: MFCD10568175
CAS Number: 1053655-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR