3-Amino-3-(2-nitrophenyl)propionic acid ≥95%
Supplier: Matrix Scientific
058736-5G
058736-25G
101915-618EA
116.92
USD
101915-618
102551-542
3-Amino-3-(2-nitrophenyl)propionic acid ≥95%
3-Amino-3-(2-nitrophenyl)propionic acid
Hazard: Irritant
Formula:
C₉H₁₀N₂O₄ |
MDL Number:
MFCD00090356 CAS Number: 5678-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...