5-Aminonicotinic acid ethyl ester ≥95%

Supplier: Matrix Scientific

058735-1G 058735-5G
101915-614EA 520 USD
101915-614 101915-616
5-Aminonicotinic acid ethyl ester ≥95%
5-Aminonicotinic acid ethyl ester

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD01087129
CAS Number: 17285-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR