N'-[1-Amino-1-m-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058733-500MG
058733-1G
101915-606EA
284.62
USD
101915-606
101915-608
N'-[1-Amino-1-m-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-m-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₉N₃O₂ MW: 249.32 g/mol |
MDL Number:
MFCD10568178 CAS Number: 1053655-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...