5-Aminomethyl-3-pyridin-3-yl[1,2,4]oxadiazole dihydrochloride ≥95%
Supplier: Matrix Scientific
058730-1G
058730-5G
101915-596EA
553.85
USD
101915-596
101915-598
5-Aminomethyl-3-pyridin-3-yl[1,2,4]oxadiazole dihydrochloride ≥95%
5-Aminomethyl-3-pyridin-3-yl[1,2,4]oxadiazole dihydrochloride
Hazard: Irritant
Formula:
C₈H₁₀Cl₂N₄O MW: 249.1 g/mol |
MDL Number:
MFCD15142769 CAS Number: 1208081-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...