5-Amino-3-methyl-isothiazole-4-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-4-cyano-3-methylisothiazole, 5-Amino-3-methylisothiazole-4-carbonitrile
058728-1G
058728-5G
101915-588EA
658.46
USD
101915-588
101915-590
5-Amino-3-methyl-isothiazole-4-carbonitrile ≥95%
5-Amino-3-methyl-isothiazole-4-carbonitrile
Hazard: Irritant
Formula:
C₅H₅N₃S MW: 139.18 g/mol |
MDL Number:
MFCD03422694 CAS Number: 41808-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...