N'-[1-Amino-1-(4-methoxyphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058727-500MG
058727-1G
101915-584EA
284.62
USD
101915-584
101915-586
N'-[1-Amino-1-(4-methoxyphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(4-methoxyphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₃H₁₉N₃O₃ MW: 265.31 g/mol |
MDL Number:
MFCD10568181 CAS Number: 1053655-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...