N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058726-500MG 058726-1G
101915-580EA 284.62 USD
101915-580 101915-582
N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₁N₃O₃
MW: 279.34 g/mol
MDL Number: MFCD10568184
CAS Number: 1053655-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR