2-Amino-5-iodonicotinic acid ≥90%
Supplier: Matrix Scientific
Synonyms:
2-Amino-5-iodopyridine-3-carboxylic acid
058724-1G
058724-5G
101915-570EA
92.31
USD
101915-570
101915-572
2-Amino-5-iodonicotinic acid ≥90%
2-Amino-5-iodonicotinic acid
Hazard: Irritant
Formula:
C₆H₅IN₂O₂ MW: 264.02 g/mol |
MDL Number:
MFCD10568356 CAS Number: 54400-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...