N'-[1-Amino-2-(2,6-dichlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058722-500MG 058722-1G
101915-562EA 284.62 USD
101915-562 101915-564
N'-[1-Amino-2-(2,6-dichlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(2,6-dichlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₇Cl₂N₃O₂
MW: 318.21 g/mol
MDL Number: MFCD10568194
CAS Number: 1053656-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR