N'-[1-Amino-2-(3,5-dichlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058721-500MG 058721-1G
101915-558EA 284.62 USD
101915-558 101915-560
N'-[1-Amino-2-(3,5-dichlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(3,5-dichlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₇Cl₂N₃O₃
MW: 334.2 g/mol
MDL Number: MFCD10568196
CAS Number: 1053656-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR