2-Amino-5-chloropyridine-3-sulfonic acid amide ≥95%
Supplier: Matrix Scientific
058717-1G
058717-5G
101915-544EA
510.77
USD
101915-544
101915-546
2-Amino-5-chloropyridine-3-sulfonic acid amide ≥95%
2-Amino-5-chloropyridine-3-sulfonic acid amide
Hazard: Irritant
Formula:
C₅H₆ClN₃O₂S |
MDL Number:
MFCD03086211 CAS Number: 163137-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...