N'-[1-Amino-1-(4-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058716-500MG
058716-1G
101915-540EA
284.62
USD
101915-540
101915-542
N'-[1-Amino-1-(4-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(4-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₂H₁₆ClN₃O₂ MW: 269.73 g/mol |
MDL Number:
MFCD10568197 CAS Number: 1053656-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...