N'-[1-Amino-1-(2-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058715-500MG
058715-1G
101915-536EA
284.62
USD
101915-536
101915-538
N'-[1-Amino-1-(2-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(2-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Hazard: Irritant
Formula:
C₁₂H₁₆ClN₃O₂ MW: 269.73 g/mol |
MDL Number:
MFCD10568187 CAS Number: 1053655-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...