N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058714-500MG 058714-1G
101915-532EA 284.62 USD
101915-532 101915-534
N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O₂
MW: 283.76 g/mol
MDL Number: MFCD10568180
CAS Number: 159016-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR